Få mere at vide
N-(Benzoyl)piperazine-2,2,3,3,5,5,6,6-d8, CDN
Artikelnummer. 30321080
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321080 0.25 g -
30309892 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321080 Leverandør CDN Leverandørnr. CDND62850.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Benzoylpiperazine-D8 (piperazine-D8)
CAS 1219805-49-8
Molekylær formel C11 D8 H6 N2 O
Mængde 0.25 g
Synonym N-(Benzoyl)piperazine-2,2,3,3,5,5,6,6-d8, 1-(Benzoyl)piperazine-2,2,3,3,5,5,6,6-d8, N-Benzoylpiperazine-d8, N-Benzoylpiperazine-D8 (piperazine-D8), Phenyl-1-(piperazinyl-d8)methanone
InChI formel InChI=1 S/C11H14N2O/c14-11(10-4-2-1-3-5-10)13-8-6-12-7-9-13/h1-5,12 H,6-9H2/i6D2,7D2,8D2,9D2
SMIL [2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])N(C(=O)c2ccccc2)C1([2 H])[2 H]
IUPAC navn (2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-phenylmethanone
Molekylvægt (g/mol) 198.291
Formel vægt 198.161 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity