missing translation for 'onlineSavingsMsg'
Få mere at vide

Thermo Scientific Chemicals N-Benzoyl-L-tyrosine ethyl ester, 98%

Artikelnummer. 10134570 Shop alle Thermo Scientific Chemicals produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
Emballage:
Glass bottle
Pakningsstørrelse:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Emballage Pakningsstørrelse
10134570 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10134570 Leverandør Thermo Scientific Chemicals Leverandørnr. 227560050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 3483-82-7 | C18H19NO4 | 313.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-Benzoyl-L-tyrosine ethyl ester
Navn note 98%
CAS 3483-82-7
CAS min % 97.5
CAS Max % 100.0
Smeltepunkt 122.3°C to 123.3°C
Farve White to Yellow
Infrarødt spektrum Authentic
Analyseprocentområde 97.5% min. (HPLC)
Emballage Glass bottle
Molekylær formel C18H19NO4
Lineær formel 4-(HO)C6H4CH2CH(NHCOC6H5)CO2C2H5
MDL nummer MFCD00002388
Mængde 5 g
Beilstein 14,I,669
Specifik rotation − 24.00
Synonym n-benzoyl-l-tyrosine ethyl ester, bz-tyr-oet, ethyl n-benzoyl-l-tyrosinate, btee, ethyl benzoyltyrosinate, benzoyltyrosine ethyl ester, benzoyl-l-tyrosine ethyl ester, s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate, benzoyl-tyr-oet, l-tyrosine, n-benzoyl-, ethyl ester
InChI nøgle SRLROPAFMUDDRC-XISACWJONA-N
SMIL CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1
IUPAC navn ethyl (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoate
Molekylvægt (g/mol) 313.35
PubChem CID 77033
Formel vægt 313.34
Procent renhed 98%
Fysisk form Powder
Vis mere Vis mindre
Produktidentifikator
  • N-Benzoyl-L-tyrosine ethyl ester, 98%

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.