missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7034620
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
51549-39-4
C23H31N5O4Si
469.62
MFCD04972281
BJZZQJFYKQPUGO-TWAUYLRJNA-N
N-{9-[(2R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl]-9H-purin-6-yl}benzamide
CC(C)(C)[Si](C)(C)OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12
This item is not returnable. View return policy
51549-39-4
C23H31N5O4Si
469.62
MFCD04972281
BJZZQJFYKQPUGO-TWAUYLRJNA-N
N-{9-[(2R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl]-9H-purin-6-yl}benzamide
CC(C)(C)[Si](C)(C)OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12

N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoyl-5′-O-tert-butyldimethylsilyl-2′-deoxyadenosine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 469.62
Percent Purity ≥98%
Chemical Name or Material N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'