Få mere at vide
N'-(Aminoiminomethyl)-N,N-dimethylurea, TRC
Artikelnummer. 30135366
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135366 250 mg
30010900 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135366 Leverandør Toronto Research Chemicals Leverandørnr. TRCA611025250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 118632-64-7
Molekylær formel C4 H10 N4 O
Molekylvægt (g/mol) 130.15
Synonym N-(N,N-Dimethylcarbamoyl)guanidine
IUPAC navn 3-carbamimidoyl-1,1-dimethylurea
SMIL CN(C)C(NC(N)=N)=O
Tekniske data
Kemisk navn eller materiale N'-(Aminoiminomethyl)-N,N-dimethylurea
Mængde 250 mg
InChI formel InChI=1S/C4H10N4O/c1-8(2)4(9)7-3(5)6/h1-2H3,(H4,5,6,7,9)
Formel vægt 130.0855