missing translation for 'onlineSavingsMsg'
Få mere at vide

N-(Aminoethyl)-5-naphthylamine-1-sulfonic Acid, TRC

Artikelnummer. 30312933
Change view
Click to view available options
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30312933 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312933 Supplier Toronto Research Chemicals Supplier No. TRCA60920025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-(Aminoethyl)-5-naphthylamine-1-sulfonic Acid
CAS 50402-56-7
Molekylær formel C12 H14 N2 O3 S
Mængde 25 g
InChI formel InChI=1S/C12H14N2O3S/c13-7-8-14-11-5-1-4-10-9(11)3-2-6-12(10)18(15,16)17/h1-6,14H,7-8,13H2,(H,15,16,17)
SMIL NCCNc1cccc2c(cccc12)S(=O)(=O)O
IUPAC navn 5-(2-aminoethylamino)naphthalene-1-sulfonic acid
Molekylvægt (g/mol) 266.32
Formel vægt 266.0725
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.