missing translation for 'onlineSavingsMsg'
Learn More

N-Amidinothiourea, 98+%, Thermo Scientific™

Catalog Number p-7022364
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
2114-02-5
C2H6N4S
118.16
MFCD00014472
OKGXJRGLYVRVNE-UHFFFAOYSA-N
This item is not returnable. View return policy
2114-02-5
C2H6N4S
118.16
MFCD00014472
OKGXJRGLYVRVNE-UHFFFAOYSA-N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Imino-4-thiobiuret is used in the design and synthesis of guanylthiourea derivatives which are potential inhibitors of plasmodium falciparum dihydrofolate reductase enzyme. 2-Imino-4-thiobiuret is also used as a reagent in recyclization reactions of N-arylmaleimides with polynucleophilic compounds.

Solubility
Soluble in water (70g/L).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 160°C to 168°C
Odor Odorless
Quantity 25 g
Beilstein 1756620
Synonym 4-pentyn-2-ol, 4-pentyne-2-ol, +--4-pentyn-2-ol, r,s-4-pentyn-2-ol, 4-hydroxypentyne, pentyne-4-ol, 4-pentin-2-ol, 1-methyl-3-butyn-1-ol, +/--4-pentyn-2-ol
Solubility Information Soluble in water (70g/L).
SMILES NC(N)=NC(N)=S
IUPAC Name (diaminomethylidene)thiourea
PubChem CID 92915
Formula Weight 118.16
Percent Purity ≥98%
Chemical Name or Material N-Amidinothiourea
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'