missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-alpha-Benzyloxycarbonyl-N-epsilon-Boc-L-lysine, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in DMF (0.5 mmol/ml).
Notes
Store at -20°C. Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in DMF (0.5 mmol/ml).
Notes
Store at -20°C. Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
Melting Point | 64°C to 67°C |
Solubility Information | Soluble in DMF (0.5 mmol/ml). |
Formula Weight | 380.44 |
Percent Purity | 99% |
Quantity | 1 g |
Chemical Name or Material | N-alpha-Benzyloxycarbonyl-N-epsilon-Boc-L-lysine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'