missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Allylphthalimide, 97%

Brand:  Alfa Aesar™ H63724.03

Additional Details : CAS Number : 5428-09-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15490559

  • 2132.98 DKK / 1g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

5428-09-1
154.25
VQUXVWMAXIQKTQ-UHFFFAOYNA-N
225150
CCCCCCC(C)(C#C)O
C10H18O
MFCD00158662
3-methyl-1-nonyn-3-ol, 3-methyl-1-nonyne-3-ol, 4,6'-sulfonyldiphenol, 3-methyl-l-nonyn-3-ol, acmc-20an14, 1-nonyn-3-ol, 3-methyl, 3-methyl-1-nonyn-3-ol #, ethynyl n-hexyl methyl carbinol, 3-methyl-1-nonyn-3-ol 5g
3-methylnon-1-yn-3-ol

Specifications

N-Allylphthalimide
1g
C10H18O
3-methyl-1-nonyn-3-ol, 3-methyl-1-nonyne-3-ol, 4,6'-sulfonyldiphenol, 3-methyl-l-nonyn-3-ol, acmc-20an14, 1-nonyn-3-ol, 3-methyl, 3-methyl-1-nonyn-3-ol #, ethynyl n-hexyl methyl carbinol, 3-methyl-1-nonyn-3-ol 5g
CCCCCCC(C)(C#C)O
154.25
187.19
97%
White
5428-09-1
MFCD00158662
VQUXVWMAXIQKTQ-UHFFFAOYNA-N
3-methylnon-1-yn-3-ol
225150
Solid
Videos
SDS Documents