Få mere at vide
N-Acetylpiperazine-d4, TRC
Artikelnummer. 30325200
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30325200 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325200 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18724710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Acetylpiperazine-3,3,5,5-D4
CAS 1398065-90-1
Molekylær formel C6 D4 H8 N2 O
Mængde 10 mg
Synonym N-Acetylpiperazine-3,3,5,5-d4, 1-Acetylpiperazine-3,3,5,5-D4, 1-(1-Piperazinyl-3,3,5,5-d4)ethanone
InChI formel InChI=1S/C6H12N2O/c1-6(9)8-4-2-7-3-5-8/h7H,2-5H2,1H3/i2D2,3D2
SMIL [2H]C1([2H])CN(CC([2H])([2H])N1)C(=O)C
IUPAC navn 1-(3,3,5,5-tetradeuteriopiperazin-1-yl)ethanone
Molekylvægt (g/mol) 132.197
Formel vægt 132.12