Få mere at vide
N-Acetylpiperazine-3,3,5,5-d4, CDN
Artikelnummer. 30319061
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319061 0.25 g -
30308847 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319061 Leverandør CDN Leverandørnr. CDND75270.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Acetylpiperazine-3,3,5,5-D4
CAS 1398065-90-1
Molekylær formel C6 D4 H8 N2 O
Mængde 0.25 g
Synonym N-Acetylpiperazine-3,3,5,5-d4, 1-Acetylpiperazine-3,3,5,5-D4, 1-(1-Piperazinyl-3,3,5,5-d4)ethanone
InChI formel InChI=1 S/C6H12N2O/c1-6(9)8-4-2-7-3-5-8/h7H,2-5H2,1H3/i2D2,3D2
SMIL [2 H]C1([2 H])CN(CC([2 H])([2 H])N1)C(=O)C
IUPAC navn 1-(3,3,5,5-tetradeuteriopiperazin-1-yl)ethanone
Molekylvægt (g/mol) 132.197
Formel vægt 132.12 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity