Få mere at vide
N-Acetylglycine N-Methylamide-d8, TRC
Artikelnummer. 30322738
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30322738 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322738 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17826410MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Acetamido-N-methylacetamide-D8
CAS 1219802-57-9
Molekylær formel C5 D8 H2 N2 O2
Mængde 10 mg
Synonym N-Acetyl-d3-glycine-d2-N-methyl-d3-amide, 2-(Acetyl-d3-amino)-N-(methyl-d3)acetamide-2,2-d2, 2-(Acet-d3-amido)-N-(methyl-d3)acetamide-2,2-d2, N-(Acetyl-d3)-N'-(methyl-d3)glycinamide-d2, 2-Acetamido-N-methylacetamide-d8, 2-Acetamido-N-methylacetamide-D8
InChI formel InChI=1S/C5H10N2O2/c1-4(8)7-3-5(9)6-2/h3H2,1-2H3,(H,6,9)(H,7,8)/i1D3,2D3,3D2
SMIL [2H]C([2H])([2H])NC(=O)C([2H])([2H])NC(=O)C([2H])([2H])[2H]
IUPAC navn 2,2,2-trideuterio-N-[1,1-dideuterio-2-oxo-2-(trideuteriomethylamino)ethyl]acetamide
Molekylvægt (g/mol) 138.194
Formel vægt 138.124