Få mere at vide
N-Acetylglycine-d5, TRC
Artikelnummer. 30328611
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30328611 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328611 Leverandør Toronto Research Chemicals Leverandørnr. TRCA178261100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetylglycine-D5 (N-acetyl,2,2-D5)
CAS 1219805-82-9
Molekylær formel C4 D5 H2 N O3
Mængde 100 mg
Synonym N-Acetylglycine-d5, N-Acetyl-d3-glycine-2,2-d2, N-Acetylglycine-D5 (N-acetyl,2,2-D5), N-Acetylglycine-d5, 2-(Acet-d3-amido)acetic-2,2-d2 acid
InChI formel InChI=1S/C4H7NO3/c1-3(6)5-2-4(7)8/h2H2,1H3,(H,5,6)(H,7,8)/i1D3,2D2
SMIL [2H]C([2H])([2H])C(=O)NC([2H])([2H])C(=O)O
IUPAC navn 2,2-dideuterio-2-[(2,2,2-trideuterioacetyl)amino]acetic acid
Molekylvægt (g/mol) 122.134
Formel vægt 122.074