Få mere at vide
N-Acetylcysteine Amide, TRC
Artikelnummer. 17983465
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17983465 50 mg
17912475 100 mg
17972775 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17983465 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17219550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38520-57-9
Molekylær formel C5H10N2O2S
Molekylvægt (g/mol) 162.21
Synonym (2R)-2-(Acetylamino)-3-mercaptopropanamide; N-Acetyl-L-cysteinamide
IUPAC navn (2R)-2-acetamido-3-sulfanylpropanamide
SMIL CC(=O)N[C@@H](CS)C(=O)N
Tekniske data
Kemisk navn eller materiale N-Acetylcysteine Amide
Renhedsgrad noter HPLC
InChI formel InChI=1S/C5H10N2O2S/c1-3(8)7-4(2-10)5(6)9/h4,10H,2H2,1H3,(H2,6,9)(H,7,8)/t4-/m0/s1
Mængde 50 mg
Procent renhed >95%