missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ N-Acetylcysteine amide

Beskrivelse
N-Acetylcysteine amide is a glutathione (GSH) precursor and cell-permeable antioxidant. N-Acetylcysteine amide replenishes intracellular Glutathione (GSH). N-Acetylcysteine amide directly reduces intracellular Glutathione disulfide (GSSG) to GSH without glutathione peroxidase. N-Acetylcysteine amide has anti-inflammatory activity through regulation of activation of NF-κB and HIF-1α, as well as modulation of reactive oxygen species. N-Acetylcysteine amide improves neuronal mitochondrial bioenergetics, reduces tissue damage and enhances functional recovery following spinal cord injury in rats. N-Acetylcysteine amide also enhances behavioral recovery in rats following traumatic brain injury. Neuroprotective.
For more information about how N-Acetylcysteine amide may be used, see our protocol: 3D Culture of Lung Alveolar Cells
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2R)-2-(Acetylamino)-3-mercaptopropanamide |
| CAS | 38520-57-9 |
| Mængde | 50 mg |
| Mål | Additional Antioxidant Compounds |
| Molekylær formel | C5H10N2O2S |
| Formel vægt | 162.21 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?