Få mere at vide
Tocris Bioscience™ N-Acetylcysteine amide
Glutathione (GSH) precursor; neuroprotective
Brand: Tocris Bioscience™ 5619/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
N-Acetylcysteine amide is a glutathione (GSH) precursor and cell permeable antioxidant. Improves neuronal mitochondrial bioenergetics, reduces tissue damage and enhances functional recovery following spinal cord injury in rats. Also enhances behavioral recovery in rats following traumatic brain injury. Neuroprotective.Tekniske data
| (2R)-2-(Acetylamino)-3-mercaptopropanamide | |
| 38520-57-9 | |
| 50 mg |