Få mere at vide
N-Acetyl Tizanidine-d4, TRC
Artikelnummer. 30320554
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30320554 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320554 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1888721MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl Tizanidine-d4
CAS 1276602-33-5
Molekylær formel C11H6D4ClN5OS
Mængde 1 mg
Synonym 1-[2-[(6-Chloro-2,1,3-benzothiadiazol-7-yl)amino]-4,4,5,5-tetradeuterioimidazol-1-yl]ethanone, 1-[2-[(5-Chloro-2,1,3-benzothiadiazol-4-yl)amino]-4,5-dihydro-d2-1H-imidazol-1-yl-4,5-d2]ethanone, 1-[2-[(5-Chloro-2,1,3-benzothiadiazol-4-yl)amino]-4,5-dihydroimidazol-1-yl-4,4,5,5-d4]ethanone, N-Acetyl tizanidine-d4, N-Acetyltizanidine-d4, N-Acetyltizanidine-D4 (dihydroimidazole-4,4,5,5-D4)
InChI formel InChI=1S/C11H10ClN5OS/c1-6(18)17-5-4-13-11(17)14-9-7(12)2-3-8-10(9)16-19-15-8/h2-3H,4-5H2,1H3,(H,13,14)/i4D2,5D2
SMIL [2H]C1([2H])N=C(Nc2c(Cl)ccc3nsnc23)N(C(=O)C)C1([2H])[2H]
IUPAC navn 1-[2-[(5-chloro-2,1,3-benzothiadiazol-4-yl)amino]-4,4,5,5-tetradeuterioimidazol-1-yl]ethanone
Molekylvægt (g/mol) 299.77