Få mere at vide
N-Acetyl Sulfapyridine-d4 (Major), TRC
Artikelnummer. 30320120
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30320120 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320120 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18791210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl Sulfapyridine-d4
CAS 1189732-52-2
Molekylær formel C13 H9 D4 N3 O3 S
Mængde 10 mg
Synonym N-[4-[(2-Pyridinyl-d4-amino)sulfonyl]phenyl]acetamide, 4’-(2-Pyridylsulfamoyl)acetanilide-d5, N4-Acetylsulfapyridine-d4
InChI formel InChI=1S/C13H13N3O3S/c1-10(17)15-11-5-7-12(8-6-11)20(18,19)16-13-4-2-3-9-14-13/h2-9H,1H3,(H,14,16)(H,15,17)/i5D,6D,7D,8D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)C)S(=O)(=O)Nc2ccccn2
IUPAC navn N-[2,3,5,6-tetradeuterio-4-(pyridin-2-ylsulfamoyl)phenyl]acetamide
Molekylvægt (g/mol) 295.35
Formel vægt 295.0929