Få mere at vide
N-Acetyl Sulfamethoxazole-d4 (major), TRC
Artikelnummer. 30313250
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30313250 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313250 Leverandør Toronto Research Chemicals Leverandørnr. TRCA187886100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylsulfamethoxazole D4
CAS 1215530-54-3
Renhedsgrad noter HPLC
Molekylær formel C12 2H4 H9 N3 O4 S
Mængde 100 mg
Synonym Acetylsulfamethoxazole D4
InChI formel InChI=1S/C12H13N3O4S/c1-8-7-12(14-19-8)15-20(17,18)11-5-3-10(4-6-11)13-9(2)16/h3-7H,1-2H3,(H,13,16)(H,14,15)/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)C)S(=O)(=O)Nc2cc(C)on2
IUPAC navn N-[2,3,5,6-tetradeuterio-4-[(5-methyl-1,2-oxazol-3-yl)sulfamoyl]phenyl]acetamide
Molekylvægt (g/mol) 299.34
Formel vægt 299.0878
Procent renhed >95