Få mere at vide
N-Acetyl Sulfadiazine-d4 (Major), TRC
Artikelnummer. 30325077
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30325077 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325077 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1878121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl Sulfadiazine-d5
CAS 1219149-66-2
Molekylær formel C12H8D4N4O3S
Mængde 1 mg
Synonym N-Acetyl Sulfadiazine D4
InChI formel InChI=1S/C12H12N4O3S/c1-9(17)15-10-3-5-11(6-4-10)20(18,19)16-12-13-7-2-8-14-12/h2-8H,1H3,(H,15,17)(H,13,14,16)/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)C)S(=O)(=O)Nc2ncccn2
IUPAC navn N-[2,3,5,6-tetradeuterio-4-(pyrimidin-2-ylsulfamoyl)phenyl]acetamide
Molekylvægt (g/mol) 296.34