Få mere at vide
N-Acetyl-S-(propyl-d7)-L-cysteine, TRC
Artikelnummer. 30324061
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30324061 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324061 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1680372.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-S-(propyl-d7)-L-cysteine
CAS 1331909-69-3
Molekylær formel C8 D7 H8 N O3 S
Mængde 2.5 mg
Synonym N-Acetyl-S-(n-propyl-d7)-L-cysteine, N-Acetyl-S-(propyl-d7)-L-cysteine, N-Acetyl-S-propyl-L-cysteine-d7, N-Acetyl-S-propyl-L-cysteine-D7 (S-propyl-D7), N-Acetyl-3-(propyl-d7-thio)-L-alanine, (R)-2-Acetamido-3-(propyl-d7-thio)propanoic acid, Propyl-d7-mercapturic acid, (2R)-2-Acetamido-3-(propylsulfanyl-d7)propanoic acid
InChI formel InChI=1S/C8H15NO3S/c1-3-4-13-5-7(8(11)12)9-6(2)10/h7H,3-5H2,1-2H3,(H,9,10)(H,11,12)/t7-/m0/s1/i1D3,3D2,4D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])SC[C@H](NC(=O)C)C(=O)O
IUPAC navn (2R)-2-acetamido-3-(1,1,2,2,3,3,3-heptadeuteriopropylsulfanyl)propanoic acid
Molekylvægt (g/mol) 212.318
Formel vægt 212.121