Få mere at vide
N-Acetyl-S-(carbamoylethyl)-L-cysteine, TRC
Artikelnummer. 17944275
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17944275 1 g
17940985 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17944275 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1718701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81690-92-8
Molekylær formel C8 H14 N2 O4 S
Molekylvægt (g/mol) 234.27
Synonym L-Cysteine, N-acetyl-S-(3-amino-3-oxopropyl)-, N-Acetyl-S-(3-amino-3-oxopropyl)-L-cysteine, (R)-2-Acetamido-3-((3-amino-3-oxopropyl)thio)propanoic acid, N-Acetyl-S-(2-carbamoylethyl)-L-cysteine
IUPAC navn (2R)-2-acetamido-3-(3-amino-3-oxopropyl)sulfanylpropanoic acid
SMIL CC(=O)N[C@@H](CSCCC(=O)N)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Acetyl-S-(carbamoylethyl)-L-cysteine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C8H14N2O4S/c1-5(11)10-6(8(13)14)4-15-3-2-7(9)12/h6H,2-4H2,1H3,(H2,9,12)(H,10,11)(H,13,14)/t6-/m0/s1
Mængde 1 g
Formel vægt 234.0674
Procent renhed >95%