Få mere at vide
N-Acetyl-S-(carbamoylethyl)-L-cysteine-d3, TRC
Artikelnummer. 30328143
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30328143 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328143 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1718725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-S-(carbamoylethyl)-L-cysteine-d3
CAS 1795786-57-0
Renhedsgrad noter HPLC
Molekylær formel C8H11D3N2O4S
Mængde 5 mg
Synonym N-(Acetyl-d3)-S-(3-amino-3-oxopropyl)-L-cysteine, AAMA-d3
InChI formel InChI=1S/C8H14N2O4S/c1-5(11)10-6(8(13)14)4-15-3-2-7(9)12/h6H,2-4H2,1H3,(H2,9,12)(H,10,11)(H,13,14)/t6-/m0/s1/i1D3
SMIL NC(CCSC[C@H](NC(C([2H])([2H])[2H])=O)C(O)=O)=O
IUPAC navn (2R)-3-(3-amino-3-oxopropyl)sulfanyl-2-[(2,2,2-trideuterioacetyl)amino]propanoic acid
Molekylvægt (g/mol) 237.29
Formel vægt 237.0863
Procent renhed >95