Få mere at vide
N-Acetyl-S-allyl-L-cysteine, TRC
Artikelnummer. 17981615
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17981615 25 mg
17983635 5 mg
17941055 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17981615 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16810025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23127-41-5
Molekylær formel C8 H13 N O3 S
Molekylvægt (g/mol) 203.26
Synonym N-Acetyl-S-2-propen-1-yl-L-cysteine, L-N-Acetyl-3-(allylthio)alanine, Allylmercapturic Acid, S-Allylmercapturic Acid
IUPAC navn (2R)-2-acetamido-3-allylsulfanyl-propanoic acid
SMIL CC(=O)N[C@@H](CSCC=C)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Acetyl-S-allyl-L-cysteine
InChI formel InChI=1S/C8H13NO3S/c1-3-4-13-5-7(8(11)12)9-6(2)10/h3,7H,1,4-5H2,2H3,(H,9,10)(H,11,12)/t7-/m0/s1
Mængde 25 mg
Formel vægt 203.0616