Få mere at vide
N-Acetyl-S-(3,4-dimethylbenzene)-L-cysteine-d3, TRC
Artikelnummer. 30325241
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30325241 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325241 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17375710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-S-(3,4-dimethylbenzene)-L-cysteine-d3
CAS 1331892-61-5
Molekylær formel C13H14D3NO3S
Mængde 10 mg
Synonym N-Acetyl-S-(3,4-dimethylphenyl)-L-cysteine-d3, 3,4-DPMA-d3
InChI formel InChI=1S/C13H17NO3S/c1-8-4-5-11(6-9(8)2)18-7-12(13(16)17)14-10(3)15/h4-6,12H,7H2,1-3H3,(H,14,15)(H,16,17)/t12-/m0/s1/i3D3
SMIL CC(C(C)=C1)=CC=C1SC[C@H](NC(C([2H])([2H])[2H])=O)C(O)=O
IUPAC navn (2R)-3-(3,4-dimethylphenyl)sulfanyl-2-[(2,2,2-trideuterioacetyl)amino]propanoic acid
Molekylvægt (g/mol) 270.36