Learn More
N-Acetyl-S-(2-hydroxypropyl)cysteine Dicyclohexylammonium Salt, TRC
Product Code. 17983155
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983155 10 mg 10mg
30007964 25 mg 25mg
17963585 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17983155 Supplier Toronto Research Chemicals Supplier No. TRCA17906010MG
 Restricted Item
Forventet forsendelse: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 38130-86-8
Molecular Formula C12 H23 N . C8 H15 N O4 S
Molecular Weight (g/mol) 402.59
Synonym L-Cysteine, N-acetyl-S-(2-hydroxypropyl)-, compd. with N-cyclohexylcyclohexanamine (1:1), N-Acetyl-S-(2-hydroxypropyl)-L-cysteine dicyclohexylamine salt, 2-Hydroxypropylmercapturic acid dicyclohexylamine salt, N-Acetyl-S-(2-hydroxypropyl)cysteine dicyclohexylamine, (2R)-2-Acetamido-3-(2-hydroxypropyl)sulfanylpropanoic acid dicyclohexylamine salt
IUPAC Name (2R)-2-acetamido-3-(2-hydroxypropylsulfanyl)propanoic acid;N-cyclohexylcyclohexanamine
SMILES CC(O)CSC[C@H](NC(=O)C)C(=O)O.C1CCC(CC1)NC2CCCCC2
Specifications
InChI Formula InChI=1S/C12H23N.C8H15NO4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(10)3-14-4-7(8(12)13)9-6(2)11/h11-13H,1-10H2;5,7,10H,3-4H2,1-2H3,(H,9,11)(H,12,13)/t;5?,7-/m.0/s1
Quantity 10 mg
Formula Weight 402.2552
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-Acetyl-S-(2-hydroxypropyl)cysteine dicyclohexylammonium salt
Grade HPLC