missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
N-Acetyl-S-(2-hydroxypropyl)cysteine-d3 Dicyclohexylammonium Salt, TRC
Specifications
Specifications
| Kemisk navn eller materiale | N-Acetyl-S-(2-hydroxypropyl)cysteine-d3 Dicyclohexylammonium salt |
| CAS | 1356934-59-2 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C12 H23 N . C8 2H3 H12 N O4 S |
| Mængde | 5 mg |
| Synonym | (2R)-3-(2-Hydroxypropylsulfanyl)-2-[(2,2,2-trideuterioacetyl)amino]propanoic acid N-Cyclohexylcyclohexanamine, N-Acetyl-S-(2-hydroxypropyl)-L-cysteine-D3 dicyclohexylamine salt (N-acetyl-D3), N-(Acetyl-d3)-S-(2-hydroxypropyl)-L-cysteine dicyclohexylamine salt, 2-Hydroxypropylmercapturic acid-d3 dicyclohexylamine salt, N-(Acetyl-d3)-S-(2-hydroxypropyl)cysteine dicyclohexylamine, (2R)-2-(Acetamido-2,2,2-d3)-3-(2-hydroxypropyl)sulfanylpropanoic acid dicyclohexylamine salt |
| InChI formel | InChI=1S/C12H23N.C8H15NO4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(10)3-14-4-7(8(12)13)9-6(2)11/h11-13H,1-10H2;5,7,10H,3-4H2,1-2H3,(H,9,11)(H,12,13)/t;5?,7-/m.0/s1/i;2D3 |
| SMIL | [2H]C([2H])([2H])C(=O)N[C@@H](CSCC(C)O)C(=O)O.C1CCC(CC1)NC2CCCCC2 |
| IUPAC navn | N-cyclohexylcyclohexanamine;(2R)-3-(2-hydroxypropylsulfanyl)-2-[(2,2,2-trideuterioacetyl)amino]propanoic acid |
| Molekylvægt (g/mol) | 405.61 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?