missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Acetyl-S-(2-hydroxyethyl-d4)-L-cysteine Dicyclohexylamine Salt, TRC

Artikelnummer. 30313372
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313372 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313372 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17891225MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-Acetyl-S-(2-hydroxyethyl-d4)-L-cysteine dicyclohexylamine salt
CAS 1331894-57-5
Renhedsgrad noter HPLC
Molekylær formel C12 H23 N . C7 2H4 H9 N O4 S
Mængde 25 mg
Synonym (2R)-2-Acetamido-3-(1,1,2,2-tetradeuterio-2-hydroxyethyl)sulfanylpropanoic acid N-cyclohexylcyclohexanamine, N-Acetyl-3-[(2-hydroxyethyl-1,1,2,2-d4)thio]-L-alanine dicyclohexylammonium salt, N-Acetyl-S-(2-hydroxyethyl-1,1,2,2-d4)-L-cysteine dicyclohexylammonium salt, N-Acetyl-S-(2-hydroxyethyl)-L-cysteine-D4 (2-hydroxyethyl-1,1,2,2-D4) dicyclohexylammonium salt, N-Acetyl-S-(2-hydroxyethyl)cysteine-d4 dicyclohexylammonium salt, S-(2-Hydroxyethyl-d4)-N-acetylcysteine dicyclohexylammonium salt, S-(β-Hydroxyethyl)mercapturic-d4 acid dicyclohexylammonium salt, (2R)-2-Acetamido-3-(2-hydroxyethyl-1,1,2,2-d4)sulfanylpropanoic acid dicyclohexylamine
InChI formel InChI=1S/C12H23N.C7H13NO4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(10)8-6(7(11)12)4-13-3-2-9/h11-13H,1-10H2;6,9H,2-4H2,1H3,(H,8,10)(H,11,12)/t;6-/m.0/s1/i;2D2,3D2
SMIL [2H]C([2H])(O)C([2H])([2H])SC[C@H](NC(=O)C)C(=O)O.C1CCC(CC1)NC2CCCCC2
IUPAC navn (2R)-2-acetamido-3-(1,1,2,2-tetradeuterio-2-hydroxyethyl)sulfanylpropanoic acid;N-cyclohexylcyclohexanamine
Molekylvægt (g/mol) 392.59
Formel vægt 392.2647
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.