missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Acetyl-S-(2-hydroxyethyl-d4)-L-cysteine Dicyclohexylamine Salt, CDN

Artikelnummer. 30324742
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324742 0.05 g -
30317951 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324742 Leverandør CDN Leverandørnr. CDND77360.05G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-Acetyl-S-(2-hydroxyethyl-d4)-L-cysteine dicyclohexylamine salt
CAS 1331894-57-5
Molekylær formel C12 H23 N . C7 2H4 H9 N O4 S
Mængde 0.05 g
Synonym (2 R)-2-Acetamido-3-(1,1,2,2-tetradeuterio-2-hydroxyethyl)sulfanylpropanoic acid N-cyclohexylcyclohexanamine, N-Acetyl-3-[(2-hydroxyethyl-1,1,2,2-d4)thio]-L-alanine dicyclohexylammonium salt, N-Acetyl-S-(2-hydroxyethyl-1,1,2,2-d4)-L-cysteine dicyclohexylammonium salt, N-Acetyl-S-(2-hydroxyethyl)-L-cysteine-D4 (2-hydroxyethyl-1,1,2,2-D4) dicyclohexylammonium salt, N-Acetyl-S-(2-hydroxyethyl)cysteine-d4 dicyclohexylammonium salt, (2 R)-2-Acetamido-3-(2-hydroxyethyl-1,1,2,2-d4)sulfanylpropanoic acid dicyclohexylamine, (2 R)-2-Acetamido-3-(2-hydroxyethyl-1,1,2,2-d4)sulfanylpropanoic acid dicyclohexylamine, (2 R)-2-Acetamido-3-(2-hydroxyethyl-1,1,2,2-d4)sulfanylpropanoic acid dicyclohexylamine
InChI formel InChI=1 S/C12H23N.C7H13NO4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(10)8-6(7(11)12)4-13-3-2-9/h11-13 H,1-10H2;6,9 H,2-4H2,1H3,(H,8,10)(H,11,12)/t;6-/m0.0/s1/i;2D2,3D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])SC[C@H](NC(=O)C)C(=O)O.C1CCC(CC1)NC2CCCCC2
IUPAC navn (2 R)-2-acetamido-3-(1,1,2,2-tetradeuterio-2-hydroxyethyl)sulfanylpropanoic acid;N-cyclohexylcyclohexanamine
Molekylvægt (g/mol) 392.59
Formel vægt 392.2647 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.