missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-S-(2,5-dimethylbenzene)-L-cysteine, TRC

Product Code. 30327401
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30327401 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327401 Supplier Toronto Research Chemicals Supplier No. TRCA17377050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Acetyl-S-(2,5-dimethylbenzene)-L-cysteine
CAS 581076-70-2
Molekylær formel C13 H17 N O3 S
Mængde 50 mg
Synonym N-Acetyl-S-(2,5-dimethylphenyl)-L-cysteine, 2,5-DPMA
InChI formel InChI=1S/C13H17NO3S/c1-8-4-5-9(2)12(6-8)18-7-11(13(16)17)14-10(3)15/h4-6,11H,7H2,1-3H3,(H,14,15)(H,16,17)/t11-/m0/s1
SMIL CC(=O)N[C@@H](CSc1cc(C)ccc1C)C(=O)O
IUPAC navn (2R)-2-acetamido-3-(2,5-dimethylphenyl)sulfanylpropanoic acid
Molekylvægt (g/mol) 267.34
Formel vægt 267.0929
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.