Få mere at vide
N-Acetyl-S-(2,2-dichloroethenyl)-L-cysteine-13C,d3, TRC
Artikelnummer. 30314337
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30314337 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314337 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1735821MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-S-(2,2-dichloroethenyl)-L-cysteine-13C,d3
CAS 1795787-09-5
Molekylær formel C613CH6D3Cl2NO3S
Mængde 1 mg
Synonym N-Acetyl-S-(2,2-dichlorovinyl)-L-cysteine-13C,d3, 2,2-DCVMA-13C,d3, NA-2,2-DCVC-13C,d3
InChI formel InChI=1S/C7H9Cl2NO3S/c1-4(11)10-5(7(12)13)2-14-3-6(8)9/h3,5H,2H2,1H3,(H,10,11)(H,12,13)/t5-/m0/s1/i1D3,4+1
SMIL O=[13C](C([2H])([2H])[2H])N[C@H](C(O)=O)CS/C=C(Cl)/Cl
IUPAC navn (2R)-3-(2,2-dichloroethenylsulfanyl)-2-[(2,2,2-trideuterioacetyl)amino]propanoic acid
Molekylvægt (g/mol) 262.13