Få mere at vide
N-Acetyl-S-(1,2-dichloroethenyl)-L-cysteine, TRC
Artikelnummer. 30134628
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134628 10 mg
30012337 5 mg
30003526 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134628 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17358510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2148-31-4
Molekylær formel C7H9Cl2NO3S
Molekylvægt (g/mol) 258.12
Synonym N-Acetyl-3-[(1,2-dichlorovinyl)thio]alanine, S-(1,2-dichlorovinyl)-L-cysteine, 1,2-DCVMA, NA-1,2-DCVC;
IUPAC navn (2R)-2-acetamido-3-[(Z)-1,2-dichloroethenyl]sulfanylpropanoic acid
SMIL Cl/C=C(Cl)/SC[C@H](NC(C)=O)C(O)=O
Tekniske data
Kemisk navn eller materiale N-Acetyl-S-(1,2-dichloroethenyl)-L-cysteine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C7H9Cl2NO3S/c1-4(11)10-5(7(12)13)3-14-6(9)2-8/h2,5H,3H2,1H3,(H,10,11)(H,12,13)/b6-2+/t5-/m0/s1
Mængde 10 mg
Formel vægt 256.968
Procent renhed >95%