Learn More
N-Acetyl-N’-phenylurea, TRC
Product Code. 30325925
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30325925 1 g 1g
1 options
Product Code. 30325925 Supplier Toronto Research Chemicals Supplier No. TRCA1923051G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phenylureidoacetic Acid
CAS 102-03-4
Molecular Formula C9 H10 N2 O2
Quantity 1 g
Synonym N-Acetyl-N'-phenylurea, Urea, 1-acetyl-3-phenyl- (7CI,8CI), N-[(Phenylamino)carbonyl]acetamide, 1-Acetyl-3-phenylurea, N-Acetyl-N'-phenylurea, NSC 60262, Phenylureidoacetic acid
InChI Formula InChI=1S/C9H10N2O2/c1-7(12)10-9(13)11-8-5-3-2-4-6-8/h2-6H,1H3,(H2,10,11,12,13)
SMILES CC(=O)NC(=O)Nc1ccccc1
IUPAC Name N-(phenylcarbamoyl)acetamide
Molecular Weight (g/mol) 178.188
Formula Weight 178.074