missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Acetyl Mesalazine, TRC

Artikelnummer. 30322896
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322896 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30322896 Leverandør Toronto Research Chemicals Leverandørnr. TRCA168320100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-Acetyl Mesalazine
CAS 51-59-2
Renhedsgrad noter HPLC
Molekylær formel C9 H9 N O4
Mængde 100 mg
Synonym 5-Acetaminosalicylic Acid
InChI formel InChI=1S/C9H9NO4/c1-5(11)10-6-2-3-8(12)7(4-6)9(13)14/h2-4,12H,1H3,(H,10,11)(H,13,14)
SMIL CC(=O)Nc1ccc(O)c(c1)C(=O)O
IUPAC navn 5-acetamido-2-hydroxybenzoic acid
Molekylvægt (g/mol) 195.17
Formel vægt 195.0532
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.