Få mere at vide
N-Acetyl Mesalazine-d3, TRC
Artikelnummer. 30313617
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30313617 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313617 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1683231MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl Mesalazine phenyl-d3
CAS 1309935-89-4
Molekylær formel C9 2H3 H6 N O4
Mængde 1 mg
Synonym 5-(Acetylamino)-2-hydroxybenzoic-d3 Acid, 3-Carboxyparacetamol-d3, 5-Acetamidosalicylic Acid-d3, CJ 46A-d3, N-Acetylmesalamine-d3, N-(Acetyl)-5-aminosalicylic Acid-d3, NSC 54183-d3
InChI formel InChI=1S/C9H9NO4/c1-5(11)10-6-2-3-8(12)7(4-6)9(13)14/h2-4,12H,1H3,(H,10,11)(H,13,14)/i2D,3D,4D
SMIL [2H]c1c([2H])c(NC(=O)C)c([2H])c(C(=O)O)c1O
IUPAC navn 3-acetamido-2,4,5-trideuterio-6-hydroxybenzoic acid
Molekylvægt (g/mol) 198.19
Formel vægt 198.072