Få mere at vide
N-Acetyl Mesalazine-d3, TRC
Artikelnummer. 30316552
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30316552 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316552 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16832225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl Mesalazine methyl-d3
CAS 93968-79-7
Molekylær formel C9 2H3 H6 N O4
Mængde 25 mg
Synonym 5-(Acetylamino-d3)-2-hydroxybenzoic Acid, 3-Carboxyparacetamol-d3, 5-Acetamidosalicylic Acid-d3, CJ 46A-d3, N-Acetylmesalamine-d3, N-(Acetyl-d3)-5-aminosalicylic Acid, NSC 54183-d3
InChI formel InChI=1S/C9H9NO4/c1-5(11)10-6-2-3-8(12)7(4-6)9(13)14/h2-4,12H,1H3,(H,10,11)(H,13,14)/i1D3
SMIL [2H]C([2H])([2H])C(=O)Nc1ccc(O)c(c1)C(=O)O
IUPAC navn 2-hydroxy-5-[(2,2,2-trideuterioacetyl)amino]benzoic acid
Molekylvægt (g/mol) 198.19
Formel vægt 198.072