Få mere at vide
N-Acetyl-L-serine-2,3,3-d3, TRC
Artikelnummer. 30319310
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30319310 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319310 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1922341MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-L-serine-2,3,3-d3
CAS 2230887-17-7
Molekylær formel C5 D3 H6 N O4
Mængde 1 mg
Synonym N-Acetyl-L-serine-2,3,3-d3
InChI formel InChI=1S/C5H9NO4/c1-3(8)6-4(2-7)5(9)10/h4,7H,2H2,1H3,(H,6,8)(H,9,10)/t4-/m0/s1/i2D2,4D
SMIL [2H]C([2H])(O)[C@]([2H])(NC(=O)C)C(=O)O
IUPAC navn (2S)-2-acetamido-2,3,3-trideuterio-3-hydroxypropanoic acid
Molekylvægt (g/mol) 150.1478
Formel vægt 150.072