Få mere at vide
N-Acetyl-L-serine-2,3,3-d3, CDN
Artikelnummer. 30322497
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322497 0.01 g
30325515 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322497 Leverandør CDN Leverandørnr. CDND69990.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-L-serine-2,3,3-d3
CAS 2230887-17-7
Molekylær formel C5 D3 H6 N O4
Mængde 0.01 g
Synonym N-Acetyl-L-serine-2,3,3-d3, N-Acetyl-L-serine-α,β,β-D3, N-Acetyl-L-serine-d3, (2 S)-2-Acetamido-3-hydroxypropanoic-2,3,3-d3 acid
InChI formel InChI=1 S/C5H9NO4/c1-3(8)6-4(2-7)5(9)10/h4,7 H,2H2,1H3,(H,6,8)(H,9,10)/t4-/m0/s1/i2D2,4 D
SMIL [2 H]C([2 H])(O)[C@]([2 H])(NC(=O)C)C(=O)O
IUPAC navn (2 S)-2-acetamido-2,3,3-trideuterio-3-hydroxypropanoic acid
Molekylvægt (g/mol) 150.1478
Formel vægt 150.072 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity