Få mere at vide
N-Acetyl-L-leucine-N-methylamide-d16, TRC
Artikelnummer. 30315183
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315183 10 mg
30316451 1 mg
30315115 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315183 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18502710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-L-leucine Methylamide D16
CAS 1276197-56-8
Molekylær formel C9 D16 H2 N2 O2
Mængde 10 mg
Synonym N-Acetyl-d3-L-leucine-d10-N-methyl-d3-amide, N-Acetyl-d3-L-leucine-d10 N-Methyl-d3-amide, (2S)-2-(Acetyl-d3-amino)-N,4-di(methyl-d3)pentanamide-2,3,3,4,5,5,5-d7
InChI formel InChI=1S/C9H18N2O2/c1-6(2)5-8(9(13)10-4)11-7(3)12/h6,8H,5H2,1-4H3,(H,10,13)(H,11,12)/t8-/m0/s1/i1D3,2D3,3D3,4D3,5D2,6D,8D
SMIL [2H]C([2H])([2H])NC(=O)[C@@]([2H])(NC(=O)C([2H])([2H])[2H])C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn (2S)-2,3,3,4,5,5,5-heptadeuterio-2-[(2,2,2-trideuterioacetyl)amino]-N,4-bis(trideuteriomethyl)pentanamide
Molekylvægt (g/mol) 202.35
Formel vægt 202.2373