Få mere at vide
N-Acetyl-L-histidine Methylamide, TRC
Artikelnummer. 30179439
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179439 100 mg
30178981 250 mg
30179157 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179439 Leverandør Toronto Research Chemicals Leverandørnr. TRCA170700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6367-11-9
Molekylær formel C9H14N4O2
Molekylvægt (g/mol) 210.23
Synonym (alphaS)-alpha-(Acetylamino)-N-methyl-1H-imidazole-5-propanamide, L-alpha-Acetamido-N-methyl-Imidazole-4-propionamide, N-Acetyl-N'-methylhistidylamide;Ac-L-His-NHMe;
IUPAC navn (2S)-2-acetamido-3-(1H-imidazol-4-yl)-N-methylpropanamide
SMIL CNC(=O)[C@H](Cc1c[nH]cn1)NC(=O)C
Tekniske data
Kemisk navn eller materiale N-Acetyl-L-histidine Methylamide
InChI formel InChI=1S/C9H14N4O2/c1-6(14)13-8(9(15)10-2)3-7-4-11-5-12-7/h4-5,8H,3H2,1-2H3,(H,10,15)(H,11,12)(H,13,14)/t8-/m0/s1
Mængde 100 mg
Formel vægt 210.1117