Få mere at vide
N-Acetyl-L-cysteine Methyl Ester, TRC
Artikelnummer. 30328714
Skift visning
Mængde:
500 g
Artikelnummer. Mængde
30328714 500 g
1 options
Artikelnummer. 30328714 Leverandør Toronto Research Chemicals Leverandørnr. TRCA145225500G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylcysteine
CAS 7652-46-2
Molekylær formel C6 H11 N O3 S
Mængde 500 g
Synonym N-Acetyl-L-cysteine methyl ester, Cysteine, N-acetyl-, methyl ester, L- (6CI,7CI,8CI), N-Acetyl-L-cysteine methyl ester, N-Acetylcysteine methyl ester, Nα-Acetyl-L-cysteine methyl ester
InChI formel InChI=1S/C6H11NO3S/c1-4(8)7-5(3-11)6(9)10-2/h5,11H,3H2,1-2H3,(H,7,8)/t5-/m0/s1
SMIL COC(=O)[C@H](CS)NC(=O)C
IUPAC navn methyl (2R)-2-acetamido-3-sulfanylpropanoate
Molekylvægt (g/mol) 177.22
Formel vægt 177.046