Få mere at vide
N-Acetyl-L-cysteine Methyl Ester, Mikromol™
Beskrivelse
N-Acetyl-L-cysteine Methyl Ester
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acetylcysteine |
| CAS | 7652-46-2 |
| Analyt- eller komponentnavne | Acetylcysteine |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C6 H11 N O3 S |
| Certificeringer/compliance | ISO 9001 |
| Synonym | N-Acetyl-L-cysteine methyl ester, Cysteine, N-acetyl-, methyl ester, L- (6CI,7CI,8CI), N-Acetylcysteine methyl ester, Nalpha-Acetyl-L-cysteine methyl ester |
| InChI formel | InChI=1S/C6H11NO3S/c1-4(8)7-5(3-11)6(9)10-2/h5,11H,3H2,1-2H3,(H,7,8)/t5-/m0/s1 |
| SMIL | COC(=O)[C@H](CS)NC(=O)C |
| IUPAC navn | methyl (2R)-2-acetamido-3-sulfanylpropanoate |
| Vis mere |