Få mere at vide
N-Acetyl-L-cysteine Isopropyl Ester, Mikromol™
Beskrivelse
N-Acetyl-L-cysteine Isopropyl Ester
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Acetyl-L-cysteine Isopropyl Ester |
| CAS | 73255-51-3 |
| Analyt- eller komponentnavne | N-Acetyl-L-cysteine Isopropyl Ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C8 H15 N O3 S |
| Certificeringer/compliance | ISO 9001 |
| Synonym | N-Acetyl-L-cysteine Isopropyl Ester, N-Acetyl-L-cysteine 1-methylethyl ester, Cysteine, N-acetyl-, isopropyl ester (6CI), N-Acetyl-L-cysteine isopropyl ester |
| InChI formel | InChI=1S/C8H15NO3S/c1-5(2)12-8(11)7(4-13)9-6(3)10/h5,7,13H,4H2,1-3H3,(H,9,10)/t7-/m0/s1 |
| SMIL | CC(C)OC(=O)[C@H](CS)NC(=O)C |
| IUPAC navn | propan-2-yl (2R)-2-acetamido-3-sulfanylpropanoate |
| Vis mere |