Få mere at vide
N-Acetyl-L-aspartic Acid, TRC
Artikelnummer. 30137335
Skift visning
Mængde:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137335 25 g
30136962 100 g
30008779 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137335 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16860525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 997-55-7
Molekylær formel C6H9NO5
Molekylvægt (g/mol) 175.14
Synonym Acetyl-L-aspartic Acid, Acetylaspartic Acid, L-N-Acetylaspartic Acid, N-Acetyl-L-aspartic Acid, N- Acetyl-S-aspartic Acid, N-Acetylaspartic Acid
IUPAC navn (2S)-2-acetamidobutanedioic acid
SMIL CC(=O)N[C@@H](CC(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Acetyl-L-aspartic Acid
Renhedsgrad noter HPLC
InChI formel InChI=1S/C6H9NO5/c1-3(8)7-4(6(11)12)2-5(9)10/h4H,2H2,1H3,(H,7,8)(H,9,10)(H,11,12)/t4-/m0/s1
Mængde 25 g
Formel vægt 175.0481
Procent renhed >95%