Få mere at vide
N-Acetyl-L-arginine Methyl Ester Hydrochloride, TRC
Artikelnummer. 30311933
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30311933 100 mg
1 options
Artikelnummer. 30311933 Leverandør Toronto Research Chemicals Leverandørnr. TRCA168360100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-L-arginine Methyl Ester Hydrochloride
CAS 1784-05-0
Molekylær formel C9 H18 N4 O3 . Cl H
Mængde 100 mg
Synonym N-Acetyl-L-arginine Methyl Ester Hydrochloride, N2-Acetyl-L-arginine Methyl Ester Monohydrochloride, Ac-L-Arg-OMe Hydrochloride
InChI formel InChI=1S/C9H18N4O3.ClH/c1-6(14)13-7(8(15)16-2)4-3-5-12-9(10)11;/h7H,3-5H2,1-2H3,(H,13,14)(H4,10,11,12);1H/t7-;/m0./s1
SMIL Cl.COC(=O)[C@H](CCCNC(=N)N)NC(=O)C
IUPAC navn methyl (2S)-2-acetamido-5-carbamimidamidopentanoate;hydrochloride
Molekylvægt (g/mol) 266.725
Formel vægt 266.115