Få mere at vide
N-Acetyl-L-alanine-3,3,3-d3, TRC
Artikelnummer. 30179369
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179369 5 mg
30179244 25 mg
30179109 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179369 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1680665MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1485548-36-4
Molekylær formel C5 2H3 H6 N O3
Molekylvægt (g/mol) 134.15
Synonym N-Acetyl-L-alanine-3,3,3-D3, N-Acetyl-L-alanine-d3, (2S)-2-Acetamido-3,3,3-trideuteriopropanoic acid, (S)-N-Acetylalanine-d3, Acetyl-L-alanine-3,3,3-d3, (S)-2-Acetamidopropionic-3,3,3-d3 acid
IUPAC navn (2S)-2-acetamido-3,3,3-trideuteriopropanoic acid
SMIL [2H]C([2H])([2H])[C@H](NC(=O)C)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Acetyl-L-alanine-3,3,3-D3
Mængde 5 mg
InChI formel InChI=1S/C5H9NO3/c1-3(5(8)9)6-4(2)7/h3H,1-2H3,(H,6,7)(H,8,9)/t3-/m0/s1/i1D3
Formel vægt 134.0771