Learn More
Alfa Aesar™ N-Acetyl-Ile-Glu-Pro-Asp-7-amino-4-(trifluoromethyl)coumarin
A fluorogenic substrate for granzyme B
Brand:  Alfa Aesar™ J65828.MA
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 15470477
  • 1789.51 DKK / 10mg
Estimated Shipment date
from Supplier 20-02-2019
Add to basket
Specifications
Specifications
10mg
−30° to −10°C
A fluorogenic substrate for granzyme B
725.66g/mol
Signal Transduction Reagent
White
Powder
C32H38N5O11F3
Ac-IEPD-AFC