Få mere at vide
N-Acetyl Glyphosate-d3, TRC
Artikelnummer. 30328329
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30328329 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328329 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17824825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl Glyphosate-d3
CAS 1346604-36-1
Renhedsgrad noter HPLC
Molekylær formel C5H7D3NO6P
Mængde 25 mg
Synonym N-(Acetyl-d3)-N-(phosphonomethyl)glycine, ( N-Acetylglyphosate-d3)
InChI formel InChI=1S/C5H10NO6P/c1-4(7)6(2-5(8)9)3-13(10,11)12/h2-3H2,1H3,(H,8,9)(H2,10,11,12)/i1D3
SMIL CC(=O)N(CC(=O)O)CP(=O)(O)O
IUPAC navn 2-[phosphonomethyl-(2,2,2-trideuterioacetyl)amino]acetic acid
Molekylvægt (g/mol) 214.13
Formel vægt 214.043
Procent renhed >95