Få mere at vide
N-Acetyl Glufosinate-d3 Disodium Salt, TRC
Artikelnummer. 30323577
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30323577 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323577 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1782375MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl Glufosinate-d3 Sodium
CAS 1356933-74-8
Molekylær formel C7H9D3NNa2O5P
Mængde 5 mg
Synonym 2-(Acetyl-d3-amino)-4-(hydroxymethylphosphinyl)butanoic acid disodium salt, 4-[Hydroxy(methyl)phosphoryl]-2-trideuterioacetamidobutanoic acid disodium salt, N-(Acetyl-d3) glufosinate disodium, N-(Acetyl-d3)-DL-glufosinate disodium, Glufosinate-N-acetyl-d3 disodium salt, Glufosinate-N-acetyl-D3 (acetyl-D3) disodium, N-Acetyl glufosinate-d3 disodium salt
InChI formel InChI=1S/C7H14NO5P.2Na/c1-5(9)8-6(7(10)11)3-4-14(2,12)13;;/h6H,3-4H2,1-2H3,(H,8,9)(H,10,11)(H,12,13);;/q;2*+1/p-2/i1D3;;
SMIL [Na+].[Na+].[2H]C([2H])([2H])C(=O)NC(CCP(=O)(C)[O-])C(=O)[O-]
IUPAC navn disodium;4-[methyl(oxido)phosphoryl]-2-[(2,2,2-trideuterioacetyl)amino]butanoate
Molekylvægt (g/mol) 270.15
Formel vægt 270.0437