Få mere at vide
N-Acetyl Dorzolamide, TRC
Artikelnummer. 30136872
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30136872 25 mg -
30133044 10 mg -
17974095 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136872 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17364025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 403848-09-9
Molekylær formel C12 H18 N2 O5 S3
Molekylvægt (g/mol) 366.4767
Synonym Acetamide, N-[(4S,6S)-2-(aminosulfonyl)-5,6-dihydro-6-methyl-7,7-dioxido-4H-thieno[2,3-b]thiopyran-4-yl]-N-ethyl-, (4S,6S)-4-[Acetyl(ethyl)amino]-6-methyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulphonamide 7,7-Dioxide, N-[(4S,6S)-2-(Aminosulfonyl)-5,6-dihydro-6-methyl-7,7-dioxido-4H-thieno[2,3-b]thiopyran-4-yl]-N-ethyl-acetamide
IUPAC navn N-ethyl-N-[(4S,6S)-6-methyl-7,7-dioxo-2-sulfamoyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-yl]acetamide
SMIL CCN([C@H]1C[C@H](C)S(=O)(=O)c2sc(cc12)S(=O)(=O)N)C(=O)C
Tekniske data
Kemisk navn eller materiale N-Acetyl Dorzolamide
Analyt- eller komponentnavne N-Acetyl Dorzolamide
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C12H18N2O5S3/c1-4-14(8(3)15)10-5-7(2)21(16,17)12-9(10)6-11(20-12)22(13,18)19/h6-7,10H,4-5H2,1-3H3,(H2,13,18,19)/t7-,10-/m0/s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 25 mg
Formel vægt 366.0378