Få mere at vide
N-Acetyl Desethyl Chloroquine-d4, TRC
Artikelnummer. 30311614
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30311614 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311614 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1735421MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl Desethyl Chloroquine 1,1,2,2-pentyl-d4
CAS 1216619-20-3
Molekylær formel C18 D4 H20 Cl N3 O
Mængde 1 mg
Synonym N-[4-[(7-Chloro-4-quinolinyl)amino]pentyl]-N-ethyl-acetamide-d4, N-Acetyl(mono)desthylchloroquine
InChI formel InChI=1S/C18H24ClN3O/c1-4-22(14(3)23)11-5-6-13(2)21-17-9-10-20-18-12-15(19)7-8-16(17)18/h7-10,12-13H,4-6,11H2,1-3H3,(H,20,21)/i5D2,11D2
SMIL [2H]C([2H])(CC(C)Nc1ccnc2cc(Cl)ccc12)C([2H])([2H])N(CC)C(=O)C
IUPAC navn N-[4-[(7-chloro-4-quinolyl)amino]-1,1,2,2-tetradeuterio-pentyl]-N-ethyl-acetamide
Molekylvægt (g/mol) 337.88
Formel vægt 337.1859